Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Chloro-1-butanol

Catalog No. 27882350ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-882-350ML 50 mL
27-882-3250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 27-882-350ML Supplier Sigma Aldrich Supplier No. 27882350ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 4-chloro-1-butanol, an alkylating agent, is one of the genotoxic impurity in active pharmaceutical ingredients
  • Reaction of 4-chloro-1-butanol and reichardtαs dye has been monitored spectrophotometrically for the determination of genotoxic impurities in pharmaceuticals.4-chloro-1-butanol was employed as an internal standard in the analysis of genotoxic impurities in active pharmaceutical ingredients by GC-ms technique.contains tetrahydrofuran and hcl

Specifications

CAS 928-51-8
Density 1.088 g/mL (at 25°C (literature))
Boiling Point 84°C to 85°C (16 mmHg)
Molecular Formula C4H9ClO
Refractive Index n20/D 1.453 (literature)
Linear Formula HO(CH2)4 Cl
MDL Number MFCD00002967
Quantity 50 mL
Synonym Tetramethylene chlorohydrin
Molecular Weight (g/mol) 108.57
Percent Purity ∼85%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.