Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-1-butanol, tech. 85%

Catalog No. AAL0404230
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0404230 250 g
AAL0404218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0404230 Supplier Thermo Scientific Chemicals Supplier No. L0404230
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 928-51-8 | C4H9ClO | 108.57 g/mol

4-Chloro-1-butanol is employed as an internal standard in the analysis of genotoxic impurities in active pharmaceutical ingredients by GC-MS technique.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloro-1-butanol is employed as an internal standard in the analysis of genotoxic impurities in active pharmaceutical ingredients by GC-MS technique.

Solubility
Soluble in methanol, chloroform, and dimethyl sufoxide.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 928-51-8
Molecular Formula C4H9ClO
Molecular Weight (g/mol) 108.57
MDL Number MFCD00002967
InChI Key HXHGULXINZUGJX-UHFFFAOYSA-N
Synonym 4-chloro-1-butanol, 4-chlorobutanol, 1-butanol, 4-chloro, tetramethylene chlorohydrin, 4-chloro-1-butane-ol, 4-chlorbutan-1-ol, unii-42ei3i5ay0, 4-chlorbutan-1-ol german, ccris 7507, 4-chloro-butan-1-ol
PubChem CID 13569
ChEBI CHEBI:87310
IUPAC Name 4-chlorobutan-1-ol
SMILES OCCCCCl

Specifications

Density 1.089
Boiling Point 50°C to 52°C (1 mmHg)
Flash Point 36°C (97°F)
Refractive Index 1.457
Linear Formula Cl(CH2)4OH
Quantity 250 g
Beilstein 1731408
Solubility Information Soluble in methanol,chloroform,and dimethyl sufoxide.
Formula Weight 108.57
Percent Purity 85%
Grade Technical
Chemical Name or Material 4-Chloro-1-butanol
Show More Show Less
Hazard Category H226-H290-H302-H314-H335-H351
Hazard Statement GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P201-P202-P210-P233-P234-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P390-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 213-175-5
RTECSNumber EL1325000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.