Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-1-butene, 98%, Thermo Scientific Chemicals

Catalog No. AC460160010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460160010 1 g
AC460160050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC460160010 Supplier Thermo Scientific Chemicals Supplier No. 460160010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 927-73-1 | C4H7Cl | 90.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 927-73-1
Molecular Formula C4H7Cl
Molecular Weight (g/mol) 90.55
InChI Key WKEVRZCQFQDCIR-UHFFFAOYSA-N
Synonym 4-chloro-1-butene, 1-butene, 4-chloro, 1-but-3-enyl chloride, ksc487m9b
PubChem CID 13565
IUPAC Name 4-chlorobut-1-ene
SMILES C=CCCCl

Specifications

Density 0.9210g/mL
Flash Point −8°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Specific Gravity 0.921
Formula Weight 90.55
Percent Purity 98%
Chemical Name or Material 4-Chloro-1-butene
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Precautionary Statement GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Store in a well-ventilated place. Keep container tightly closed.
EINECSNumber 213-160-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.