Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-1-methylpiperidine 98.0+%, TCI America™
SDP

Catalog No. C24655G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24655G 5 g
C246525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C24655G Supplier TCI America Supplier No. C24655G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5570-77-4
Molecular Formula C6H12ClN
Molecular Weight (g/mol) 133.62
MDL Number MFCD00044489
InChI Key MYGXGCCFTPKWIH-UHFFFAOYSA-N
PubChem CID 79342
IUPAC Name 4-chloro-1-methylpiperidine
SMILES CN1CCC(Cl)CC1

Specifications

Color Yellow
Boiling Point 162°C
Quantity 5 g
UN Number 1993
Formula Weight 133.62
Percent Purity ≥98.0% (GC,T)
Grade Not Available
Physical Form Liquid
Chemical Name or Material 4-Chloro-1-methylpiperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.