Learn More
Alkali Scientific 4 Chloro 1 Naphthol, 100 Grams
SDP
Catalog No. 5000661908
Encompass_Preferred
4-Chloro-1-Naphthol is a biochemical reagent used in various enzyme assays and microbiological studies. This 100g pack is ideal for laboratories requiring a large quantity of this reagent for high-throughput testing. Chloro-naphthol undergoes a color change upon hydrolysis by enzymes like peroxidase, providing a clear visual indication of enzyme activity. It is frequently used in conjunction with substrates such as hydrogen peroxide to measure peroxidase activity in biological samples. 4-Chloro-1-Naphthol is an essential tool for studies in enzymology, histochemistry, and microbiology, offering a reliable method for detecting oxidative enzymes in both clinical and research environments.
Catalog No. 50-006-61908 Supplier Alkali Scientific Supplier No. RON001CP
4-Chloro-1-Naphthol is a biochemical reagent used in various enzyme assays and microbiological studies. This 100g pack is ideal for laboratories requiring a large quantity of this reagent for high-throughput testing. Chloro-naphthol undergoes a color change upon hydrolysis by enzymes like peroxidase, providing a clear visual indication of enzyme activity. It is frequently used in conjunction with substrates such as hydrogen peroxide to measure peroxidase activity in biological samples. 4-Chloro-1-Naphthol is an essential tool for studies in enzymology, histochemistry, and microbiology, offering a reliable method for detecting oxidative enzymes in both clinical and research environments.