Learn More
Alkali Scientific 4 Chloro 1 Naphthol, 25 G
SDP
Catalog No. 5000661673
Encompass_Preferred
Chloro-naphthol is a chromogenic reagent used in biochemical assays, particularly in enzyme detection and blotting procedures. The reagent reacts with alkaline phosphatase and other phosphatases to form a colored precipitate, making it an essential tool in various laboratory techniques, including immunoblotting, enzyme-linked assays, and histochemical staining. This 25g bottle is ideal for preparing multiple solutions for large-scale assays and laboratory research. It is commonly used in molecular biology and microbiology for detecting specific enzymes and proteins, helping scientists identify bacterial species and detect changes in gene expression or enzyme activity in a variety of sample types.
Catalog No. 50-006-61673 Supplier Alkali Scientific Supplier No. RON002CP
Chloro-naphthol is a chromogenic reagent used in biochemical assays, particularly in enzyme detection and blotting procedures. The reagent reacts with alkaline phosphatase and other phosphatases to form a colored precipitate, making it an essential tool in various laboratory techniques, including immunoblotting, enzyme-linked assays, and histochemical staining. This 25g bottle is ideal for preparing multiple solutions for large-scale assays and laboratory research. It is commonly used in molecular biology and microbiology for detecting specific enzymes and proteins, helping scientists identify bacterial species and detect changes in gene expression or enzyme activity in a variety of sample types.