Learn More
Alkali Scientific 4 Chloro 1 Naphthol, 500 G
SDP
Catalog No. 5000661909
Encompass_Preferred
Chloro-naphthol is a versatile chromogenic reagent used in enzyme detection and blotting applications. This 500g bulk format is ideal for large-scale laboratory work, allowing researchers to prepare ample quantities of reagent for multiple assays. It is particularly useful in immunoassays, enzyme-linked immunosorbent assays (ELISA), and Western blotting, where it produces a visible color change in the presence of alkaline phosphatase activity. Chloro-naphthol is also commonly used in microbiological research to detect specific bacterial enzymes, aiding in microbial identification and enzymatic studies. This reagent is essential for accurate and sensitive enzyme detection in a wide range of laboratory protocols.
Catalog No. 50-006-61909 Supplier Alkali Scientific Supplier No. RON003CP
Chloro-naphthol is a versatile chromogenic reagent used in enzyme detection and blotting applications. This 500g bulk format is ideal for large-scale laboratory work, allowing researchers to prepare ample quantities of reagent for multiple assays. It is particularly useful in immunoassays, enzyme-linked immunosorbent assays (ELISA), and Western blotting, where it produces a visible color change in the presence of alkaline phosphatase activity. Chloro-naphthol is also commonly used in microbiological research to detect specific bacterial enzymes, aiding in microbial identification and enzymatic studies. This reagent is essential for accurate and sensitive enzyme detection in a wide range of laboratory protocols.