Learn More
Medchemexpress LLC 4-Chloro-1-naphthol | 604-44-4 | 178.62 | 1 ML
SDP
Catalog No. 5000448625
Encompass_Preferred
4-Chloro-1-naphthol is a substrate of horseradish peroxidase. It can be used to visualize protein bands in western blotting and immunohistochemical staining, and also as a substrate to stain cryosections black by indirect immunoperoxidase staining.
  • Functions as a substrate for horseradish peroxidase
  • Useful for visualizing protein bands in western blotting
  • Applicable in immunohistochemical staining
  • Can be used to stain cryosections black via indirect immunoperoxidase staining
Catalog No. 50-004-48625 Supplier Medchemexpress LLC Supplier No. HYW00796710MM/1ML
4-Chloro-1-naphthol is a substrate of horseradish peroxidase. It can be used to visualize protein bands in western blotting and immunohistochemical staining, and also as a substrate to stain cryosections black by indirect immunoperoxidase staining.
  • Functions as a substrate for horseradish peroxidase
  • Useful for visualizing protein bands in western blotting
  • Applicable in immunohistochemical staining
  • Can be used to stain cryosections black via indirect immunoperoxidase staining