Learn More
Medchemexpress LLC 4-Chloro-1-naphthol | 604-44-4 | 178.62 | 500 MG
SDP
Catalog No. 5000447871
Encompass_Preferred
4-Chloronaphthalen-1-ol is a substrate of horseradish peroxidase. It can be used to visualize protein bands in western blotting and immunohistochemical staining.
  • Used to visualize protein bands in western blotting.
  • Used in immunohistochemical staining.
  • Substrate to stain cryosections black by indirect immunoperoxidase staining.
Catalog No. 50-004-47871 Supplier Medchemexpress LLC Supplier No. HYW007967500MG
4-Chloronaphthalen-1-ol is a substrate of horseradish peroxidase. It can be used to visualize protein bands in western blotting and immunohistochemical staining.
  • Used to visualize protein bands in western blotting.
  • Used in immunohistochemical staining.
  • Substrate to stain cryosections black by indirect immunoperoxidase staining.