Learn More
Abcam 4-Chloro-1-naphthol, Chromogenic pero x idase substrate, 10G
Catalog No. 502704601
Encompass_Preferred
MW 178.61 g/mol. Chromogenic peroxidase substrate that results in a precipitate that is blue to blue-purple in color and can be observed visually. Used for both western blotting and immunohistochemical staining. Can be used as a substitute in the myeloperoxidase stain for benzidine compounds, which are considered to be carcinogenic.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4601 Supplier Abcam Supplier No. AB14652210G
MW 178.61 g/mol. Chromogenic peroxidase substrate that results in a precipitate that is blue to blue-purple in color and can be observed visually. Used for both western blotting and immunohistochemical staining. Can be used as a substitute in the myeloperoxidase stain for benzidine compounds, which are considered to be carcinogenic.
The product is subject to the following: Abcam Restricted Use Statement