Learn More
Medchemexpress LLC 4-chloro-1-piperidin-4-yl-N-(5-pyridin-2-yl-1H-pyrazol-4-yl)pyrazole-3-carboxamide | 1817698-21-7 | MFCD28386213 | >98.0% | 371.8 g/mol | C17H18ClN7O | 50MG
SDP
Catalog No. 5000237852
Encompass_Preferred
BDP5290 is a small-molecule kinase inhibitor selective for Rho-associated kinases (ROCK) and myotonic dystrophy kinase-related Cdc42-binding kinases (MRCK). It is used as a research reagent in biochemical and cell-based assays to probe pathways controlling myosin light chain phosphorylation, cell motility, and tumor cell invasion.
  • Potent inhibition of ROCK1 (IC50 5 nM), ROCK2 (IC50 50 nM), MRCKα (IC50 10 nM), and MRCKβ (IC50 100 nM).
  • High reported purity: >98.0%.
  • Molecular formula C17H18ClN7O and molecular weight 371.8 g/mol.
  • Suitable for in vitro and cell-based research applications investigating cytoskeletal regulation.
Catalog No. 50-002-37852 Supplier Medchemexpress LLC Supplier No. HY1243750MG
Add to Cart
Add to Cart
BDP5290 is a small-molecule kinase inhibitor selective for Rho-associated kinases (ROCK) and myotonic dystrophy kinase-related Cdc42-binding kinases (MRCK). It is used as a research reagent in biochemical and cell-based assays to probe pathways controlling myosin light chain phosphorylation, cell motility, and tumor cell invasion.
  • Potent inhibition of ROCK1 (IC50 5 nM), ROCK2 (IC50 50 nM), MRCKα (IC50 10 nM), and MRCKβ (IC50 100 nM).
  • High reported purity: >98.0%.
  • Molecular formula C17H18ClN7O and molecular weight 371.8 g/mol.
  • Suitable for in vitro and cell-based research applications investigating cytoskeletal regulation.