Learn More
Sigma Aldrich 4-Chloro-1H-pyrrolo[2,3-b]pyridine-5-carbonitrile
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Ethyl linoleate can undergo auto-oxidation in the presence of the catalyst manganese(ii)acetylacetonate.ethyl linoleate can be used as a drying agent for alkyd paints
- It can also form n2,3-ethenoguanine via reaction with deoxyguanosine.ethyl linoleate solated from oxalis triangularis inhibits forskolin-induced melanogenesis and tyrosinase activity in mouse b16 melanoma cells
- This anti-melanogenic effect is mediated by inhibiting camp production
Specifications
Specifications
| CAS | 920966-02-5 |
| Molecular Formula | C8H4ClN3 |
| Linear Formula | C8H4ClN3 |
| MDL Number | MFCD09965877 |
| Quantity | 500 mL |
| Molecular Weight (g/mol) | 177.59 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.