Learn More
Selleck Chemical LLC 4-Chloro-2-(trifluoroacetyl)aniline hydrochloride S5423-25mg
SDP
Catalog No. 503110966
Encompass_Preferred
4-Chloro-2-(trifluoroacetyl)aniline hydrochloride is a HIV-1 RT (HIV reverse transcriptase) inhibitor
Catalog No. 50-311-0966 Supplier Selleck Chemical LLC Supplier No. S542325MG
4-Chloro-2-(trifluoroacetyl)aniline hydrochloride is a HIV-1 RT (HIV reverse transcriptase) inhibitor