Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-2-(1H-pyrazol-3-yl)phenol 98.0+%, TCI America™
SDP

Catalog No. C28185G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C28185G Supplier TCI America Supplier No. C28185G
Add to Cart
Add to Cart

Specifications

CAS 18704-67-1
Melting Point 159°C
Color White-Yellow
Molecular Formula C9H7ClN2O
Quantity 5 g
MDL Number MFCD00209566
Synonym 4-chloro-2-1h-pyrazol-3-yl phenol, 3-5-chloro-2-hydroxyphenyl pyrazole, phenol,4-chloro-2-1h-pyrazol-3-yl, acmc-20amm6, dmglumyoloaxjy-uhfffaoysa-n, 2-1h-pyrazole-5-yl-4-chlorophenol, 4-chloro-2-1h-pyrazol-3-yl-phenol, phenol, 4-chloro-2-1h-pyrazol-3-yl, 4-chloro-2-2h-pyrazol-3-yl phenol, 4-chloro-6-1,2-dihydropyrazol-3-ylidene-1-cyclohexa-2,4-dienone
InChI Key BAWPNSUPRVQKKN-UHFFFAOYSA-N
SMILES C1=CC(=O)C(=C2C=CNN2)C=C1Cl
IUPAC Name 4-chloro-6-(1,2-dihydropyrazol-3-ylidene)cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol) 194.618
PubChem CID 6805776
Formula Weight 194.62
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-2-(1H-pyrazol-3-yl)phenol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.