Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-2,5-dimethylaniline 98.0+%, TCI America™
SDP

Catalog No. C31455G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31455G 5 g
C31451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31455G Supplier TCI America Supplier No. C31455G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 20782-94-9 | C8H10ClN | 155.625 g/mol

Chemical Identifiers

CAS 20782-94-9
Molecular Formula C8H10ClN
Molecular Weight (g/mol) 155.625
MDL Number MFCD08445705
InChI Key POEANWWEPFQEEF-UHFFFAOYSA-N
Synonym 4-chloro-2,5-xylidine, 4-chloro-2,5-dimethylphenylamine, 4-chloro-2,5-dimethyl-phenylamine, acmc-20amm1, 4-chloranyl-2,5-dimethyl-aniline, benzenamine,4-chloro-2,5-dimethyl, 4-chloro-2,5-dimethyl-anilin
PubChem CID 12338307
IUPAC Name 4-chloro-2,5-dimethylaniline
SMILES CC1=CC(=C(C=C1Cl)C)N

Specifications

Melting Point 93°C
Color White-Yellow
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Formula Weight 155.63
Percent Purity ≥98.0% (GC,T)
Grade Not Available
Chemical Name or Material 4-Chloro-2,5-dimethylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.