Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-2,6-dimethylaniline hydrochloride, 98%, Thermo Scientific™

Catalog No. AAA1936209
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1936209 10 g
AAA1936218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1936209 Supplier Thermo Scientific Chemicals Supplier No. A1936209

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5769-32-4
Molecular Formula C8H11Cl2N
Molecular Weight (g/mol) 192.083
MDL Number MFCD00671703
InChI Key MHTIYKGRFKCWAU-UHFFFAOYSA-N
Synonym 4-chloro-2,6-dimethylaniline hydrochloride, acmc-1astt, 4-chloro-2,6-xylidine hydrochloride, 4-chloro-2,6-dimethylaniline hcl, 4-chloranyl-2,6-dimethyl-aniline hydrochloride, 4-chloro-2,6-dimethylaniline-hydrogen chloride 1/1
PubChem CID 18406981
IUPAC Name 4-chloro-2,6-dimethylaniline;hydrochloride
SMILES CC1=CC(=CC(=C1N)C)Cl.Cl

Specifications

Quantity 10 g
Formula Weight 192.09
Percent Purity 98%
Chemical Name or Material 4-Chloro-2,6-dimethylaniline hydrochloride
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.