Learn More
Medchemexpress LLC 4-chloro-2-[[(E)-3-(4-pentylphenyl)prop-2-enoyl]amino]benzoic acid | 99754-06-0 | 99.3% | 371.86 | C21H22ClNO3 | 10MG
SDP
Catalog No. 5000238642
Encompass_Preferred
ONO-RS-082 is a reversible inhibitor of phospholipase A2 (PLA2) used in biochemical and cell-based research to probe PLA2-dependent signaling. It inhibits PLA2 with an IC50 of 1.0 μM and does not inhibit phospholipase C at concentrations up to 100 μM. The compound is supplied as a high-purity small molecule with documented solubility and storage recommendations for experimental use.
  • Reversible PLA2 inhibition (IC50 = 1.0 μM)
  • No PLC inhibition up to 100 μM
  • High purity suitable for research applications
  • Soluble in DMSO; in vivo formulation protocols available
  • Stable as powder at -20°C for long-term storage
Catalog No. 50-002-38642 Supplier Medchemexpress LLC Supplier No. HY12307010MG
ONO-RS-082 is a reversible inhibitor of phospholipase A2 (PLA2) used in biochemical and cell-based research to probe PLA2-dependent signaling. It inhibits PLA2 with an IC50 of 1.0 μM and does not inhibit phospholipase C at concentrations up to 100 μM. The compound is supplied as a high-purity small molecule with documented solubility and storage recommendations for experimental use.
  • Reversible PLA2 inhibition (IC50 = 1.0 μM)
  • No PLC inhibition up to 100 μM
  • High purity suitable for research applications
  • Soluble in DMSO; in vivo formulation protocols available
  • Stable as powder at -20°C for long-term storage