Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-2-fluoro-5-nitrotoluene, 98%

Catalog No. AC426010050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC426010050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC426010050 Supplier Thermo Scientific Chemicals Supplier No. 426010050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 18349-11-6 | C7H5ClFNO2 | 189.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18349-11-6
Molecular Formula C7H5ClFNO2
Molecular Weight (g/mol) 189.57
MDL Number MFCD00134231
InChI Key SJDPAVRCQNFVDM-UHFFFAOYSA-N
Synonym 4-chloro-2-fluoro-5-nitrotoluene, 1-chloro-5-fluoro-4-methyl-2-nitro-benzene, toluene, 4-chloro-2-fluoro-5-nitro, benzene,1-chloro-5-fluoro-4-methyl-2-nitro, benzene, 1-chloro-5-fluoro-4-methyl-2-nitro, acmc-1c7av, 4-chloro-2-fluoro-5-nitro-toluene, buttpark 146\15-31, 2-chloro-4-floro-5-methylnitrobenzene, 2-chloro-4-fluoro-5-methylnitrobenzene
PubChem CID 87593
IUPAC Name 1-chloro-5-fluoro-4-methyl-2-nitrobenzene
SMILES CC1=CC(=C(Cl)C=C1F)[N+]([O-])=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 42.0°C to 45.0°C
Color Yellow to Brown
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 189.57
Percent Purity 98%
Physical Form Crystalline Mass
Chemical Name or Material 4-Chloro-2-fluoro-5-nitrotoluene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre
EINECSNumber 242-225-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.