Learn More
Medchemexpress LLC 4-chloro-2-fluorobenzeneboronic acid | 160591-91-3 | 174.37 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW002971
Encompass_Preferred
4-Chloro-2-fluorobenzeneboronic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • Biochemical reagent
  • Biological material
  • Organic compound for life science research
  • Solid appearance
  • White to off-white color
  • Soluble in DMSO (100 mg/mL)
Catalog No. 50-004-48291