Learn More
Medchemexpress LLC 4-chloro-2-fluorobenzoic acid | 446-30-0 | MFCD00042468 | 99.3% | 174.56 | C7H4ClFO2 | 50 G
SDP
Catalog No. 5000301927
Encompass_Preferred
4-Chloro-2-fluorobenzoic acid is a halogenated benzoic acid used as a biochemical reagent and building block in organic and medicinal chemistry. It is supplied as a solid with high purity and is intended for research applications such as synthesis, assay development, and structure-activity investigations.
  • High purity for reliable experimental results.
  • Suitable as an intermediate for organic synthesis.
  • Applicable in medicinal chemistry and SAR studies.
  • Available in multiple pack sizes for lab-scale use.
  • Characterization data (HNMR, RP-HPLC) and safety documents available.
Catalog No. 50-003-01927 Supplier Medchemexpress LLC Supplier No. HYW01676950G
4-Chloro-2-fluorobenzoic acid is a halogenated benzoic acid used as a biochemical reagent and building block in organic and medicinal chemistry. It is supplied as a solid with high purity and is intended for research applications such as synthesis, assay development, and structure-activity investigations.
  • High purity for reliable experimental results.
  • Suitable as an intermediate for organic synthesis.
  • Applicable in medicinal chemistry and SAR studies.
  • Available in multiple pack sizes for lab-scale use.
  • Characterization data (HNMR, RP-HPLC) and safety documents available.