Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-2-fluorobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. C16685G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16685G 5 g
C16681G 1 g
C166825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C16685G Supplier TCI America Supplier No. C16685G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 446-30-0
Molecular Formula C7H4ClFO2
Molecular Weight (g/mol) 174.555
MDL Number MFCD00042468
InChI Key ZLPXBWMVZANJJQ-UHFFFAOYSA-N
Synonym 2-fluoro-4-chlorobenzoic acid, benzoic acid, 4-chloro-2-fluoro, 4-chloro-2-fluorobenzoicacid, 4-chloro-2-fluoro-benzoic acid, pubchem1374, acmc-209jz4, ksc237m4j, rarechem al bo 0755, 4-chloro-2-fluoro benzoic acid, timtec-bb sbb003797
PubChem CID 99152
IUPAC Name 4-chloro-2-fluorobenzoic acid
SMILES C1=CC(=C(C=C1Cl)F)C(=O)O

Specifications

Melting Point 206°C
Color White
Quantity 5 g
Formula Weight 174.56
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-2-fluorobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.