Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-2-fluorophenol, 98%, Thermo Scientific™

Catalog No. AAH6141614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6141614 25 g
AAH6141606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6141614 Supplier Thermo Scientific Chemicals Supplier No. H6141614

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 348-62-9
Molecular Formula C6H4ClFO
Molecular Weight (g/mol) 146.55
MDL Number MFCD00075030
InChI Key ZKMUKBBWORLNLA-UHFFFAOYSA-N
Synonym 2-fluoro-4-chlorophenol, phenol, 4-chloro-2-fluoro, 4-chloro-2-fluoro-phenol, 4-chloro-fluorophenol, pubchem2861, acmc-1ct1y, ksc222g4r, 4-chloro-2-fluorophenol, attercop-chm at106713
PubChem CID 181975
IUPAC Name 4-chloro-2-fluorophenol
SMILES OC1=CC=C(Cl)C=C1F

Specifications

Melting Point 20°C
Color Colorless to Yellow
Density 1.378 g/mL
Boiling Point 103°C to 104°C
Flash Point 81°C (178°F)
Refractive Index 1.536
Quantity 25 g
Formula Weight 146.55
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Chloro-2-fluorophenol
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.