Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-2-fluorophenol 98.0+%, TCI America™
SDP

Catalog No. C22645G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22645G 5 g
C226425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C22645G Supplier TCI America Supplier No. C22645G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 348-62-9
Molecular Formula C6H4ClFO
Molecular Weight (g/mol) 146.55
MDL Number MFCD00075030
InChI Key ZKMUKBBWORLNLA-UHFFFAOYSA-N
Synonym 2-fluoro-4-chlorophenol, phenol, 4-chloro-2-fluoro, 4-chloro-2-fluoro-phenol, 4-chloro-fluorophenol, pubchem2861, acmc-1ct1y, ksc222g4r, 4-chloro-2-fluorophenol, attercop-chm at106713
PubChem CID 181975
IUPAC Name 4-chloro-2-fluorophenol
SMILES OC1=CC=C(Cl)C=C1F

Specifications

Melting Point 20°C
Color Yellow
Boiling Point 104°C
Quantity 5 g
Formula Weight 146.55
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Chloro-2-fluorophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.