Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-chloro-2-(furan-2-ylmethylamino)-5-sulfamoylbenzoic acid | 54-31-9 | MFCD00010549 | 100.0% | 330.74 | C12H11ClN2O5S | 10 MG
SDP

Catalog No. 5000297062
Encompass_Preferred

Furosemide reference standard is an analytical-grade reference material of the loop diuretic furosemide intended for research and analytical applications. It is provided as a 10 mg solid with high purity suitable for assay calibration, method development, and quality control in laboratories.

  • High purity: 99.95%.
  • CAS number: 54-31-9.
  • Molecular formula: C12H11ClN2O5S.
  • Molecular weight: 330.74.
  • Pack size: 10 mg solid.
  • Intended for analytical and research use as a reference standard.

Catalog No. 50-002-97062 Supplier Medchemexpress LLC Supplier No. HYB0135R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Furosemide reference standard is an analytical-grade reference material of the loop diuretic furosemide intended for research and analytical applications. It is provided as a 10 mg solid with high purity suitable for assay calibration, method development, and quality control in laboratories.

  • High purity: 99.95%.
  • CAS number: 54-31-9.
  • Molecular formula: C12H11ClN2O5S.
  • Molecular weight: 330.74.
  • Pack size: 10 mg solid.
  • Intended for analytical and research use as a reference standard.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.