Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-2-methoxyphenol, 99%, Thermo Scientific™

Catalog No. AC294110100
Change view
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC294110100 10 g Glass bottle
AC294110050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC294110100 Supplier Thermo Scientific Chemicals Supplier No. 294110100
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16766-30-6
Molecular Formula C7H7ClO2
Molecular Weight (g/mol) 158.58
MDL Number MFCD00070774
InChI Key FVZQMMMRFNURSH-UHFFFAOYSA-N
PubChem CID 28050
IUPAC Name 4-chloro-2-methoxyphenol
SMILES COC1=CC(Cl)=CC=C1O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 16.0°C to 17.0°C
Color Yellow to Brown
Density 1.3000g/mL
Boiling Point 241.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.5575 to 1.5595
Linear Formula ClC6H3(OCH3)OH
Quantity 10 g
Specific Gravity 1.3
Synonym 4-chloroguaiacol, phenol, 4-chloro-2-methoxy, 4-chloro-2-methoxy-phenol, 2-methoxy-4-chloro-phenol, pubchem3665, acmc-209dwx, #, phenol,4-chloro-2-methoxy, 4-chloranyl-2-methoxy-phenol, fvzqmmmrfnursh-uhfffaoysa
Formula Weight 158.58
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Chloro-2-methoxyphenol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.