Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-2-methylphenol, 97%, Thermo Scientific™

Catalog No. AC109475000
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC109475000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC109475000 Supplier Thermo Scientific Chemicals Supplier No. 109475000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1570-64-5
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.582
InChI Key RHPUJHQBPORFGV-UHFFFAOYSA-N
Synonym 4-chloro-o-cresol, 2-methyl-4-chlorophenol, 4-chloro-2-cresol, p-chloro-o-cresol, phenol, 4-chloro-2-methyl, o-cresol, 4-chloro, 4-chloro-2-methyl phenol, 4-chloro-2-methyl-phenol, 5-chloro-2-hydroxytoluene, unii-297v63w9ri
PubChem CID 14855
ChEBI CHEBI:1800
IUPAC Name 4-chloro-2-methylphenol
SMILES CC1=C(C=CC(=C1)Cl)O

Specifications

Melting Point 43.0°C to 46.0°C
Boiling Point 220.0°C to 225.0°C
Flash Point 103°C
Linear Formula ClC6H3(CH3)OH
Quantity 500 g
Solubility Information Solubility in water: 7.6g/L (25°C). Other solubilities: readily soluble in caustic soda solution,, aromatic solvents, ethanol and acetone, moderately soluble in paraffinic solvents and, petroleum ether
Formula Weight 142.58
Chemical Name or Material 4-Chloro-2-methylphenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.