Learn More
Medchemexpress LLC 4-chloro-2-methylphenol-d4 | 1219804-06-4 | 99.6% | 146.61 g/mol | C7H3D4ClO | 10 MG
SDP
Catalog No. 5000297891
Encompass_Preferred
4-Chloro-2-methylphenol-d4 is the deuterium-labeled analogue of 4-chloro-2-methylphenol provided for research use as an isotopic tracer and internal standard. It is intended for analytical and method-development workflows where accurate quantitation and traceability are required.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • High purity (99.6%) for accurate quantitation.
  • Molecular weight 146.61 g/mol; formula C7H3D4ClO.
  • Available in small milligram pack sizes for analytical workflows.
Catalog No. 50-002-97891 Supplier Medchemexpress LLC Supplier No. HY34498S10MG
4-Chloro-2-methylphenol-d4 is the deuterium-labeled analogue of 4-chloro-2-methylphenol provided for research use as an isotopic tracer and internal standard. It is intended for analytical and method-development workflows where accurate quantitation and traceability are required.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • High purity (99.6%) for accurate quantitation.
  • Molecular weight 146.61 g/mol; formula C7H3D4ClO.
  • Available in small milligram pack sizes for analytical workflows.