Learn More
Medchemexpress LLC 4-Chloro-2-methylphenol-d4 (4-chloro-o-cresol-d4) | 1219804-06-4 | 99.6% | 146.61 | 50 MG
SDP
Catalog No. 5000404308
Encompass_Preferred
4-Chloro-2-methylphenol-d4 is a deuterium-labeled version of 4-Chloro-2-methylphenol. It is intended for research use only and is not sold to patients. This compound can be used as a tracer and as an internal standard for quantitative analysis by methods such as NMR, GC-MS, or LC-MS. Deuterium substitution, as seen in this compound, has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Functions as an internal standard for quantitative analysis.
  • Stable heavy isotope.
Catalog No. 50-004-04308 Supplier Medchemexpress LLC Supplier No. HY34498S50MG
4-Chloro-2-methylphenol-d4 is a deuterium-labeled version of 4-Chloro-2-methylphenol. It is intended for research use only and is not sold to patients. This compound can be used as a tracer and as an internal standard for quantitative analysis by methods such as NMR, GC-MS, or LC-MS. Deuterium substitution, as seen in this compound, has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Functions as an internal standard for quantitative analysis.
  • Stable heavy isotope.