Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4-Chloro-2-methylphenoxy)acetic Acid 98.0+%, TCI America™
SDP

Catalog No. C0206500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0206500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C0206500G Supplier TCI America Supplier No. C0206500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Melting Point 119°C
CAS 94-74-6
Quantity 500 g
Physical Form Crystal-Powder at 20°C
Molecular Formula C9H9ClO3
MDL Number MFCD00004306
Synonym mcpa, agroxone, 2-methyl-4-chlorophenoxyacetic acid, 4-chloro-2-methylphenoxy acetic acid, zelan, metaxon, 2-4-chloro-2-methylphenoxy acetic acid, bordermaster, agroxon, chiptox
InChI Key WHKUVVPPKQRRBV-UHFFFAOYSA-N
SMILES CC1=C(C=CC(=C1)Cl)OCC(=O)O
IUPAC Name 2-(4-chloro-2-methylphenoxy)acetic acid
Molecular Weight (g/mol) 200.618
PubChem CID 7204
ChEBI CHEBI:50099
Formula Weight 200.62
Percent Purity ≥98.0% (T)
Chemical Name or Material (4-Chloro-2-methylphenoxy)acetic Acid
Show More Show Less
EINECSNumber (3)-0922
RTECSNumber AG1575000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.