Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-2-nitrobenzaldehyde 98.0+%, TCI America™
SDP

Catalog No. C09645G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C09645G 5 g
C096425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C09645G Supplier TCI America Supplier No. C09645G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5551-11-1
Molecular Formula C7H4ClNO3
Molecular Weight (g/mol) 185.56
MDL Number MFCD00056104
InChI Key MZPNQUMLOFWSEK-UHFFFAOYSA-N
PubChem CID 79674
IUPAC Name 4-chloro-2-nitrobenzaldehyde
SMILES C1=CC(=C(C=C1Cl)[N+](=O)[O-])C=O

Specifications

Melting Point 68°C
Quantity 5 g
Formula Weight 185.56
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-2-nitrobenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.