Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Chloro-2-(quinazolin-4-ylamino)phenol hydrochloride

Catalog No. OTV0008951G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OTV0008951G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OTV0008951G Supplier Sigma Aldrich Supplier No. OTV0008951G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Peroxynitrite promoted aerobic oxidation of 3-methyl-2,4-pentanedione has been reported
  • The kinetics of the reaction of oh radicals with 3-methyl-2,4-pentanedione has been investigated in the gas-phase using a pulsed laser photolysis-laser induced fluorescence technique
  • 3-methyl-2,4-pentanedione participates in asymmetric substitution of 1,3-diphenyl-2-propenyl acetate catalyzed by amphiphilic resin-supported monodentate phosphine ligands

Specifications

Molecular Formula C14H11Cl2N3O
Linear Formula C14H11Cl2N3O
Quantity 25 g
MDL Number MFCD03618640
Molecular Weight (g/mol) 308.16
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.