Learn More
Medchemexpress LLC 4-chloro-3-[[(3-nitrophenyl)amino]sulfonyl]-benzoic acid | 412940-35-3 | MFCD01122300 | 98.0% | 356.74 g·mol⁻¹ | C13H9ClN2O6S | 10 MG
SDP
Catalog No. 5000248615
Encompass_Preferred
CTP inhibitor (Compound 792949) is a potent, selective inhibitor of citrate transport proteins (CTP/PMCT) used in biochemical research to investigate citrate transport and related metabolic pathways. The compound is supplied at high purity as a solid and is also available as a DMSO solution for in vitro experiments. For research use only.
  • Potent, selective inhibition of citrate transport proteins.
  • High purity appropriate for biochemical assays.
  • Available in multiple milligram quantities and as a DMSO solution.
  • Useful for studies of citrate transport and metabolic pathways.
  • Provided for research use only; not for human consumption.
Catalog No. 50-002-48615 Supplier Medchemexpress LLC Supplier No. HY10199810MG
Add to Cart
Add to Cart
CTP inhibitor (Compound 792949) is a potent, selective inhibitor of citrate transport proteins (CTP/PMCT) used in biochemical research to investigate citrate transport and related metabolic pathways. The compound is supplied at high purity as a solid and is also available as a DMSO solution for in vitro experiments. For research use only.
  • Potent, selective inhibition of citrate transport proteins.
  • High purity appropriate for biochemical assays.
  • Available in multiple milligram quantities and as a DMSO solution.
  • Useful for studies of citrate transport and metabolic pathways.
  • Provided for research use only; not for human consumption.