Learn More
Medchemexpress LLC 4-chloro-3-((3-nitrophenyl)sulfamoyl)benzoic acid | 412940-35-3 | 98.0% | 356.74 g/mol | C13H9ClN2O6S | 1 ML
SDP
Catalog No. 5000248022
Encompass_Preferred
CTP inhibitor is a selective small-molecule inhibitor of the plasma membrane citrate transporter (PMCT). It is supplied as a 10 mM solution in DMSO for direct use in cellular and biochemical assays that probe citrate transport and cellular metabolism.
  • Selective inhibitor of the plasma membrane citrate transporter (PMCT).
  • Supplied as a 10 mM solution in DMSO for direct use in assays.
  • Packaged as a 1 mL ready-to-use aliquot.
  • Purity typically ≥98.0% (HPLC), suitable for research applications.
  • Supporting documentation includes data sheet, COA, SDS, HNMR, and LCMS.
Catalog No. 50-002-48022 Supplier Medchemexpress LLC Supplier No. HY10199810MM 1ML
CTP inhibitor is a selective small-molecule inhibitor of the plasma membrane citrate transporter (PMCT). It is supplied as a 10 mM solution in DMSO for direct use in cellular and biochemical assays that probe citrate transport and cellular metabolism.
  • Selective inhibitor of the plasma membrane citrate transporter (PMCT).
  • Supplied as a 10 mM solution in DMSO for direct use in assays.
  • Packaged as a 1 mL ready-to-use aliquot.
  • Purity typically ≥98.0% (HPLC), suitable for research applications.
  • Supporting documentation includes data sheet, COA, SDS, HNMR, and LCMS.