Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-3,5-dimethylphenol, MP Biomedicals

Catalog No. ICN209827 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN209827 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN209827 Supplier MP Biomedicals, Inc Supplier No. 0520982705

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 88-04-0
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.609
InChI Key OSDLLIBGSJNGJE-UHFFFAOYSA-N
Synonym chloroxylenol, dettol, 4-chloro-3,5-xylenol, pcmx, benzytol, p-chloro-m-xylenol, 2-chloro-m-xylenol, 4-chloro-m-xylenol, ottasept, desson
PubChem CID 2723
ChEBI CHEBI:34393
IUPAC Name 4-chloro-3,5-dimethylphenol
SMILES CC1=CC(=CC(=C1Cl)C)O

Specifications

Quantity 10 g
Formula Weight 156.61
Chemical Name or Material 4-Chloro-3, 5-dimethylphenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.