Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich {[(4-Chloro-3,5-dimethylphenoxy)acetyl]amino}acetic acid

Catalog No. OTV0003821G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OTV0003821G 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OTV0003821G Supplier Sigma Aldrich Supplier No. OTV0003821G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Sodium 2-ethylhexyl sulfate is an anionic surfactant that can be used:
  • In suspension polymerization
  • In the analysis of phenolic compounds through microchip-ce with pulsed amperometric detection
  • As charge balancing anions in the synthesis of organo-layered double hydroxides (organo-ldhs).alkyl sodium sulfate surfactants

Specifications

CAS 853743-94-9
Molecular Formula C12H14ClNO4
Linear Formula C12H14ClNO4
MDL Number MFCD06449735
Quantity 1 L
Molecular Weight (g/mol) 271.7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.