Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-3,5-dinitrobenzonitrile 98.0+%, TCI America™
SDP

Catalog No. C10115G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C10115G Supplier TCI America Supplier No. C10115G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1930-72-9
Molecular Formula C7H2ClN3O4
Molecular Weight (g/mol) 227.56
MDL Number MFCD00007067
InChI Key SCGDEDHSPCXGEC-UHFFFAOYSA-N
Synonym benzonitrile, 4-chloro-3,5-dinitro, 4-chloro-3,5-dinitrobenzenenitrile, 1-chloro-4-cyano-2,6-dinitrobenzene, 4-chloro-3,5-dinitrobenzenecarbonitrile, acmc-1brcu, 4-chloro-3,5-dinitrobenzonitril, 3,5-dinitro-4-chlorobenzonitrile, 4-cyano-2,6-dinitrochlorobenzene, 4-chloro-3,5-dinitro-benzonitrile, benzonitrile,4-chloro-3,5-dinitro
PubChem CID 16008
IUPAC Name 4-chloro-3,5-dinitrobenzonitrile
SMILES C1=C(C=C(C(=C1[N+](=O)[O-])Cl)[N+](=O)[O-])C#N

Specifications

Melting Point 140°C
Color Yellow
Quantity 5 g
Formula Weight 227.56
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-3,5-dinitrobenzonitrile
RTECSNumber DI2990000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.