Learn More
Medchemexpress LLC 4-chloro-3-(5-methyl-3-{[4-(2-pyrrolidin-1-ylethoxy)phenyl]amino}-1,2,4-benzotriazin-7-yl)phenyl benzo | 867331-82-6 | MFCD16038901 | 98.6% | 580.08 g/mol | C33H30ClN5O3 | 100 MG
SDP
Catalog No. 5000249000
Encompass_Preferred
TG 100801 is a prodrug that is de-esterified to generate an active multi-targeted kinase inhibitor used in research on age-related macular degeneration. The compound is characterized by CAS 867331-82-6, formula C33H30ClN5O3, and a molecular weight of 580.08 g/mol; it is supplied for laboratory research and documented in chemical databases.
  • Prodrug that converts to an active multi-targeted kinase inhibitor.
  • Relevant to ophthalmic research into age-related macular degeneration.
  • High chemical purity suitable for in vitro and preclinical studies.
  • Available in salt form for improved handling and solubility.
  • Well characterized with CAS, formula, and molecular weight for traceability.
Catalog No. 50-002-49000 Supplier Medchemexpress LLC Supplier No. HY10186100MG
TG 100801 is a prodrug that is de-esterified to generate an active multi-targeted kinase inhibitor used in research on age-related macular degeneration. The compound is characterized by CAS 867331-82-6, formula C33H30ClN5O3, and a molecular weight of 580.08 g/mol; it is supplied for laboratory research and documented in chemical databases.
  • Prodrug that converts to an active multi-targeted kinase inhibitor.
  • Relevant to ophthalmic research into age-related macular degeneration.
  • High chemical purity suitable for in vitro and preclinical studies.
  • Available in salt form for improved handling and solubility.
  • Well characterized with CAS, formula, and molecular weight for traceability.