Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-3-fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. C27495G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2749-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C2749-5G Supplier TCI America Supplier No. C27495G
Add to Cart
Add to Cart

Specifications

CAS 137504-86-0
Melting Point 144°C
Color White
Molecular Formula C6H5BClFO2
MDL Number MFCD01319010
Quantity 5 g
Synonym 4-chloro-3-fluorophenyl boronic acid, 4-chloro-3-fluorobenzeneboronic acid, 3-fluoro-4-chlorophenylboronic acid, 4-chloro-3-fluoro-phenyl boronic acid, boronic acid, 4-chloro-3-fluorophenyl
InChI Key CMJQIHGBUKZEHP-UHFFFAOYSA-N
SMILES B(C1=CC(=C(C=C1)Cl)F)(O)O
IUPAC Name (4-chloro-3-fluorophenyl)boronic acid
Molecular Weight (g/mol) 174.362
PubChem CID 2778656
Formula Weight 174.36
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.