Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-3-methylpyridine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog No. AAL1955803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1955803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1955803 Supplier Thermo Scientific Chemicals Supplier No. L1955803
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 19524-08-4 | Molecular Formula: C6H7Cl2N | Molecular Weight: 164.029 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19524-08-4
Molecular Formula C6H7Cl2N
Molecular Weight (g/mol) 164.029
MDL Number MFCD03092890
InChI Key WPIUSHWGBZMBHZ-UHFFFAOYSA-N
Synonym 4-chloro-3-methylpyridine hydrochloride, 4-chloro-3-picoline hcl, 4-chloro-3-methylpyridine hcl, 4-chloro-3-picoline hydrochloride, pyridine, 4-chloro-3-methyl-, hydrochloride, 4-chloro-3-methylpyridinehydrochloride, acmc-20amlc, pubchem9218, ksc495e6r, 4-chloro-3methylpyridine hydrochloride
PubChem CID 16217655
IUPAC Name 4-chloro-3-methylpyridine;hydrochloride
SMILES CC1=C(C=CN=C1)Cl.Cl

Specifications

Melting Point ∼185°C
Odor Odorless
Quantity 1 g
Beilstein 107953
Formula Weight 164.04
Percent Purity 97%
Chemical Name or Material 4-Chloro-3-methylpyridine hydrochloride
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.