Learn More
Medchemexpress LLC 4-Chloro-3-nitrobenzoic acid | 96-99-1 | 99.39% | 201.56 | 1 ML
SDP
Catalog No. 5000401209
Encompass_Preferred
4-Chloro-3-nitrobenzoic acid exhibits oral activity, and when administered orally, it can influence the synthesis of liver proteins as well as the activities of both urokinase and histidine kinase.
  • Has oral activity.
  • Affects the synthesis activity of liver proteins.
  • Influences the activities of urokinase.
  • Impacts the activities of histidine kinase.
Catalog No. 50-004-01209 Supplier Medchemexpress LLC Supplier No. HY2055910MM/1ML
4-Chloro-3-nitrobenzoic acid exhibits oral activity, and when administered orally, it can influence the synthesis of liver proteins as well as the activities of both urokinase and histidine kinase.
  • Has oral activity.
  • Affects the synthesis activity of liver proteins.
  • Influences the activities of urokinase.
  • Impacts the activities of histidine kinase.