Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-3-nitrobenzoyl chloride, 98%, Thermo Scientific™

Catalog No. AC235635000
Change view
Click to view available options
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC235635000 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC235635000 Supplier Thermo Scientific Chemicals Supplier No. 235635000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 38818-50-7
CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C7H3Cl2NO3
Quantity 500g
MDL Number MFCD00035743
Synonym 3-nitro-4-chlorobenzoyl chloride, benzoyl chloride, 4-chloro-3-nitro, 4-chloro-3-nitrobenzoylchloride, acmc-1afrh, ksc494s0t, timtec-bb sbb006725, 4-chloro-3-nitro-benzoylchloride, attercop-chm at106925, 4-chloro-3-nitro-benzoyl chloride, art-chem-bb b022432
InChI Key IWLGXPWQZDOMSB-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1C(=O)Cl)[N+](=O)[O-])Cl
IUPAC Name 4-chloro-3-nitrobenzoyl chloride
Molecular Weight (g/mol) 220.005
PubChem CID 123477
Formula Weight 220.02
Percent Purity ≥97.5% (RCOCL)
Chemical Name or Material 4-Chloro-3-nitrobenzoyl chloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Causes serious eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 254-133-6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.