Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-3-nitrophenyl isocyanate, 94%, Thermo Scientific™

Catalog No. AAL1108703
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1108703 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1108703 Supplier Thermo Scientific Chemicals Supplier No. L1108703

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 40397-96-4
Molecular Formula C7H3ClN2O3
Quantity 1g
MDL Number MFCD00037062
UN Number UN2811
Beilstein 2807129
Sensitivity Moisture sensitive
Synonym 4-chloro-3-nitrophenyl isocyanate, 4-chloro-3-nitrophenylisocyanate, 3-nitro-4-chlorophenyl isocyanate, 4-chloro-3-nitrobenzenisocyanate, acmc-209jdv, #, benzene,1-chloro-4-isocyanato-2-nitro, 1-chloranyl-4-isocyanato-2-nitro-benzene, isocyanic acid 4-chloro-3-nitrophenyl ester
InChI Key ZCPHLSPLEGBTCZ-UHFFFAOYSA-N
SMILES [O-][N+](=O)C1=CC(=CC=C1Cl)N=C=O
IUPAC Name 1-chloro-4-isocyanato-2-nitrobenzene
Molecular Weight (g/mol) 198.56
PubChem CID 142448
Formula Weight 198.57
Percent Purity 94%
Chemical Name or Material 4-Chloro-3-nitrophenyl isocyanate
Show More Show Less
Hazard Category H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H331-H334-H302-H312-H315-H319-H317-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.