Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-3-nitrophenyl Isocyanate 95.0+%, TCI America™
SDP

Catalog No. I05645G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I05645G Supplier TCI America Supplier No. I05645G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 40397-96-4
Molecular Formula C7H3ClN2O3
Molecular Weight (g/mol) 198.56
MDL Number MFCD00037062
InChI Key ZCPHLSPLEGBTCZ-UHFFFAOYSA-N
Synonym 4-chloro-3-nitrophenyl isocyanate, 4-chloro-3-nitrophenylisocyanate, 3-nitro-4-chlorophenyl isocyanate, 4-chloro-3-nitrobenzenisocyanate, acmc-209jdv, #, benzene,1-chloro-4-isocyanato-2-nitro, 1-chloranyl-4-isocyanato-2-nitro-benzene, isocyanic acid 4-chloro-3-nitrophenyl ester
PubChem CID 142448
IUPAC Name 1-chloro-4-isocyanato-2-nitrobenzene
SMILES [O-][N+](=O)C1=CC(=CC=C1Cl)N=C=O

Specifications

Melting Point 67°C
Color Yellow
Quantity 5 g
UN Number 2206
Formula Weight 198.56
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-3-nitrophenyl Isocyanate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.