Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-chloro-3-nitrophenyl isothiocyanate, Thermo Scientific™

Catalog No. SB01784DE
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB01784DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SB01784DE Supplier Thermo Scientific Chemicals Supplier No. SB01784DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 127142-66-9
Molecular Formula C7H3ClN2O2S
Molecular Weight (g/mol) 214.623
InChI Key ZXGZBHIDSJXKLE-UHFFFAOYSA-N
Synonym 4-chloro-3-nitrophenyl isothiocyanate, 4-chloro-3-nitrophenylisothiocyanate, acmc-1c0qb, 4-chloro-3-nitrobenzenisothiocyanate, #, benzene,1-chloro-4-isothiocyanato-2-nitro, 1-chloranyl-4-isothiocyanato-2-nitro-benzene
PubChem CID 145581
IUPAC Name 1-chloro-4-isothiocyanato-2-nitrobenzene
SMILES C1=CC(=C(C=C1N=C=S)[N+](=O)[O-])Cl

Specifications

Assay Percent Range 97%
Quantity 5 g
Formula Weight 214.63
Percent Purity 97%
Chemical Name or Material 4-Chloro-3-nitrophenyl isothiocyanate
Hazard Category GHS Signal Word: Danger
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.