Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-3-nitropyridine hydrochloride, 90+%, Thermo Scientific™

Catalog No. AAL1972803
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1972803 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1972803 Supplier Thermo Scientific Chemicals Supplier No. L1972803

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 54079-68-4
Molecular Formula C5H4Cl2N2O2
Quantity 1g
MDL Number MFCD02683026
Synonym 4-chloro-3-nitropyridine hydrochloride, 4-chloro-3-nitropyridine monohydrochloride, 4-chloro-3-nitropyridine, chloride, c5h3cln2o2.clh, acmc-20a6v1, 3-nitro-4-chloropyridine hydrochloride, 4-chloro-3-nitro-pyridine hydrochloride, 4-chloro-3-nitropyridinemonohydrochloride, 4-chloranyl-3-nitro-pyridine hydrochloride
InChI Key YTPDMOPPRVGKEW-UHFFFAOYSA-N
SMILES C1=CN=CC(=C1Cl)[N+](=O)[O-].Cl
IUPAC Name 4-chloro-3-nitropyridine;hydrochloride
Molecular Weight (g/mol) 194.999
PubChem CID 12244206
Formula Weight 195
Percent Purity ≥90%
Chemical Name or Material 4-Chloro-3-nitropyridine hydrochloride
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.