Learn More
Sigma Aldrich 4-Chloro-3-(tributylstannyl)pyridine
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Enantiodifferentiating geometrical photoisomerizations of (z,z)-1,3-cyclooctadiene was studied using cyclodextrin nanosponge, as a supramolecular sensitizing host
- Dendrimer-encapsulated pd-rh bimetallic nanoparticle catalyzed partial hydrogenation of 1,3-cyclooctadiene has been reported.1,3-cyclooctadiene has been used in the preparation of 2-cyclooct-2-en-1-yl-1,3-cyclooctadiene, a novel bicyclic triene
Specifications
Specifications
| CAS | 206115-40-4 |
| Molecular Formula | C17H30ClNSn |
| Linear Formula | C17H30ClNSn |
| Quantity | 10 mL |
| MDL Number | MFCD20483587 |
| Molecular Weight (g/mol) | 402.59 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.