Learn More
Accela Chembio Inc 4-chloro-3-(trifluoromethyl)benzoic Acid | 25g | 1737-36-6 | MFCD01631468 | 97+% | Retest Period: 540days | Light Sensitive
Catalog No. 50648839
Encompass_Preferred
4-chloro-3-(trifluoromethyl)benzoic Acid | 25g | 1737-36-6 | MFCD01631468 | 97+% | Retest Period: 540days | Light Sensitive
Catalog No. 50-648-839 Supplier Accela Chembio Inc Supplier No. SY02366925G
4-chloro-3-(trifluoromethyl)benzoic Acid | 25g | 1737-36-6 | MFCD01631468 | 97+% | Retest Period: 540days | Light Sensitive