Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-3-(trifluoromethyl)benzaldehyde 97.0+%, TCI America™
SDP

Catalog No. C27601G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27601G 1 g
C27605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C27601G Supplier TCI America Supplier No. C27601G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 34328-46-6
Molecular Formula C8H4ClF3O
Molecular Weight (g/mol) 208.564
MDL Number MFCD00040722
InChI Key NIHMMULLFBKTOK-UHFFFAOYSA-N
Synonym 4-chloro-3-trifluoromethyl benzaldehyde, 4-chloro-3-trifluoromethyl-benzaldehyde, 2-chloro-5-formylbenzotrifluoride, benzaldehyde, 4-chloro-3-trifluoromethyl, pubchem2892, acmc-1adw8, timtec-bb sbb003468, 4-chloro-3trifluoromethyl-benzaldehyde, 3-trifluoromethyl-4-chloro-benzaldehyde, 4-chloranyl-3-trifluoromethyl benzaldehyde
PubChem CID 118642
IUPAC Name 4-chloro-3-(trifluoromethyl)benzaldehyde
SMILES C1=CC(=C(C=C1C=O)C(F)(F)F)Cl

Specifications

Color Yellow
Boiling Point 54°C
Quantity 1 g
Formula Weight 208.56
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Chloro-3-(trifluoromethyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.