Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-3-(trifluoromethyl)benzenesulfonyl Chloride 98.0+%, TCI America™
SDP

Catalog No. C27055G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27055G 5 g
C27051G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C27055G Supplier TCI America Supplier No. C27055G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 32333-53-2
Molecular Formula C7H3Cl2F3O2S
Molecular Weight (g/mol) 279.054
MDL Number MFCD00024882
InChI Key SSULGNXFUGLULI-UHFFFAOYSA-N
Synonym 4-chloro-3-trifluoromethyl benzenesulfonyl chloride, 4-chloro-3-trifluoromethyl benzene-1-sulfonyl chloride, 4-chloro-3-trifluoromethyl-benzenesulfonyl chloride, 4-chloro-3-trifluoromethyl benzenesulfonylchloride, 4-chloro-3-trifluoromethyl benzenesulphonyl chloride, 4-chloro-5-trifluoromethyl benzenesulfonyl chloride, benzenesulfonyl chloride, 4-chloro-3-trifluoromethyl, ksc222e0j, 4-chloro-3-trifluoromethylbenzenesulfonylchloride, 4-chloro-3-trifluoromethyl benzenesulfonyl chlori
PubChem CID 2780719
IUPAC Name 4-chloro-3-(trifluoromethyl)benzenesulfonyl chloride
SMILES C1=CC(=C(C=C1S(=O)(=O)Cl)C(F)(F)F)Cl

Specifications

Color White
Boiling Point 131°C
Quantity 5 g
UN Number 3261
Formula Weight 279.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Chloro-3-(trifluoromethyl)benzenesulfonyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.