Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Chloro-4-dimethylaminoazobenzene 98.0+%, TCI America™
SDP

Catalog No. C015925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C015925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C015925G Supplier TCI America Supplier No. C015925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2491-76-1
Molecular Formula C14H14ClN3
Molecular Weight (g/mol) 259.737
MDL Number MFCD00059481
InChI Key WQWHNMIYCHFRJK-UHFFFAOYSA-N
Synonym 4-Dimethylamino-4′C-chloroazobenzene
PubChem CID 17226
IUPAC Name 4-[(4-chlorophenyl)diazenyl]-N,N-dimethylaniline
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)Cl

Specifications

Melting Point 159°C
Quantity 25 g
Formula Weight 259.74
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Chloro-4-dimethylaminoazobenzene
RTECSNumber BX5060000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.